Skip to Content
Merck
All Photos(1)

Key Documents

900447

Sigma-Aldrich

Red light-emitting spiro-copolymer

average Mw 470,000

Synonym(s):

Livilux® SPR-001

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$675.00

$675.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$675.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

$675.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

solid

Quality Level

mol wt

average Mn 105,000
average Mw 470,000

Looking for similar products? Visit Product Comparison Guide

Application

Features:
  • Soluble red light-emitting spiro-copolymer.
  • Long lifetime, with further big potential to improve.
  • Saturated red color coordinators.
  • Low operation voltage.
  • Very good viscosity behaviour in many organic solvents.

Typical performance measured in standard device structure (ITO/PEDOT:PSS 20 nm/SPR-001 80 nm/Ba 6 nm/Ag 150 nm):
  • Max. efficiency: 1.90 Cd/A.
  • V_on: 2.00 V.
  • V at 100 cd/m2: 3.3 V.
  • V at 1000 cd/m2: 5.1 V.
  • Max. power eff.: 1.80 lm/W.
  • External quantum eff.: 2.44%.
  • CIE1931 at 100 cd/m2: (0.67, 0.33).
  • DC lifetime at 100 cd/m2 at RT: >20,000.

Preparation Note

  • 11 g/L in toluene for spin-coating.
  • Special formulation needed for inkjet.
  • Preparation should be in N2 atmosphere.

Legal Information

Product of EMD Performance Materials Corp., an Affiliate of Merck KGaA, Darmstadt, Germany
Livilux is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service