Skip to Content
Merck
All Photos(3)

Key Documents

900432

Sigma-Aldrich

Tetra-n-butylammonium decatungstate

Synonym(s):

1-Butanaminium, N,N,N-tributyl-, eicosa-μ-oxodi-μ5-oxodecaoxodecatungstate (4:1), 1-Butanaminium, N,N,N-tributyl-, salt with tungstic acid (4:1), TBADT, Tetrakis(N,N,N-tributyl-1-butanaminium) tungstate, Tetrakis(tetrabutylammonium) decatungstate

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$173.00
1 G
$572.00

$173.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$173.00
1 G
$572.00

About This Item

Empirical Formula (Hill Notation):
C64H144N4O32W10
CAS Number:
Molecular Weight:
3320.24
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.22

$173.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

Quality Level

reaction suitability

core: tungsten
reaction type: C-H Activation
reaction type: Photocatalysis
reagent type: catalyst

Application

When used in conjunction with cobaloxime pyridine chloride (341630), this complex can effect the catalytic, room-temperature dehydrogenation of unactivated alkanes under solar irradiation with hydrogen as the sole byproduct.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

The Sorensen group has reported a dual-catalytic system capable of dehydrogenating alkanes & alcohols at low temperatures with hydrogen gas as the sole byproduct

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service