900431
SGA 545 100 isiphor
Synonym(s):
Europium doped alkaline earth orthosilicate, Green phosphor, Rare earth doped phosphor
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
powder
composition
Halogen free, Br<700 ppm, Cl <700 ppm, Br+Cl <1000 ppm
particle size
11-13 μm (D50)
17.5-19.5 μm (D90)
5.5-7.5 μm (D10)
fluorescence
λem 544 nm±1 nm
Application
Phosphors are solid inorganic crystalline materials that exhibit luminescence upon excitation. Applications of phosphors extend from bio-imaging/bio-sensors to X-ray detectors, fluorescent lamps, displays, and LEDs. Our YYG series isiphor are yttrium garnet based yellow phosphors whereas the SGA series isiphor are silicate based green phosphors. These materials exhibit >90% quantum efficiency and are well suited for application in backlighting for liquid crystal displays and LEDs.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Nano Letters, 4, 2004-2004 (2004)
Long persistent phosphors-from fundamentals to applications.
Chemical Society Reviews, 45, 2090-2090 (2016)
J. Alloy Compounds, 275, 669-676 (1998)
Nano Letters, 4, 2191-2191 (2004)
The journal of physical chemistry letters, 2(11), 1268-1277 (2011-06-02)
Light-emitting diodes (LEDs) are excellent candidates for general lighting because of their rapidly improving efficiency, durability, and reliability, their usability in products of various sizes, and their environmentally friendly constituents. Effective lighting devices can be realized by combining one or
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service