900428
SGA 524 100 isiphor
Synonym(s):
Europium doped alkaline earth orthosilicate, Green phosphor, Rare earth doped phosphor
About This Item
Recommended Products
form
powder
composition
Halogen free, Br: <700 ppm, Cl: <700 ppm, Br+Cl: <1000 ppm
particle size
14-16 μm (D50)
21.5-23.5 μm (D90)
7.5-9.5 μm (D10)
fluorescence
λem 524 nm±1 nm
Application
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service