Skip to Content
Merck
All Photos(1)

Documents

900378

Sigma-Aldrich

Jasmonic-2,4,4-d3-(acetyl-2,2-d2) acid

with variable deuteration on OD, ≥97 atom % D, ≥97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12D5H13O3
Molecular Weight:
215.30
UNSPSC Code:
12352106
PubChem Substance ID:

isotopic purity

≥97 atom % D

Assay

≥97%

form

liquid

technique(s)

mass spectrometry (MS): suitable

mass shift

M+2

storage temp.

2-8°C

SMILES string

O=C1[C@]([2H])(C/C=C\CC)[C@@H](C([2H])(C(O[R])=O)[2H])CC1([2H])[2H]

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service