Skip to Content
Merck
All Photos(1)

Key Documents

900300

Sigma-Aldrich

Poly(L-lactide-co-caprolactone)

greener alternative

lactide:caprolactone 60:40, viscosity 1.5 dL/g 

Synonym(s):

PLLA-PCL

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$358.00

$358.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$358.00

About This Item

Linear Formula:
CH3(C6H10O2)m(C6H8O4)nCH3
UNSPSC Code:
12162002
NACRES:
NA.23

$358.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

chips or granules

Quality Level

feed ratio

lactide:caprolactone 60:40

greener alternative product characteristics

Design for Energy Efficiency
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

viscosity

1.5 dL/g

greener alternative category

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which belongs to one of the four categories of greener alternatives. This is an enabling product that belongs to a family of high-performance bio-based polymers that are essential for developing efficient, low-cost, and environmentally friendly renewable energy solutions. Click here for more information.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

A Poulos et al.
Advances in experimental medicine and biology, 318, 331-340 (1992-01-01)
Fatty acids with from 24 to 28 carbon atoms (very long-chain fatty acids, VLCFA) are present in small amounts in all mammalian tissues. Even longer chain fatty acids with from 30 to 38 carbon atoms (ultra-long-chain fatty acids, ULCFA) are

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service