Skip to Content
Merck
All Photos(2)

Key Documents

900275

Sigma-Aldrich

(t-Bu)PhCPhos

95%

Synonym(s):

2-[(tert-Butyl)phenylphosphino]-2′,6′-bis(N,N-dimethylamino)biphenyl

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$263.00
1 G
$742.00

$263.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$263.00
1 G
$742.00

About This Item

Empirical Formula (Hill Notation):
C26H33N2P
CAS Number:
Molecular Weight:
404.53
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22

$263.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

powder or crystals

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

reagent type: ligand

mp

112 °C

functional group

phosphine

SMILES string

P(C(C)(C)C)(c2c(cccc2)c3c(cccc3N(C)C)N(C)C)c1ccccc1

InChI

1S/C26H33N2P/c1-26(2,3)29(20-14-9-8-10-15-20)24-19-12-11-16-21(24)25-22(27(4)5)17-13-18-23(25)28(6)7/h8-19H,1-7H3

InChI key

HTHOMNNVAUPLKK-UHFFFAOYSA-N

Application

(t-Bu)PhCPhos is the Pd G3 complex, from the Buchwald group offers best-in-class performance in the arylation of sterically hindered primary amines.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paula Ruiz-Castillo et al.
Journal of the American Chemical Society, 137(8), 3085-3092 (2015-02-05)
We report the Pd-catalyzed arylation of very hindered α,α,α-trisubstituted primary amines. Kinetics-based mechanistic analysis and rational design have led to the development of two biarylphosphine ligands that allow the transformation to proceed with excellent efficiency. The process was effective in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service