Skip to Content
Merck
All Photos(1)

Documents

900235

Sigma-Aldrich

CdSe/ZnS core-shell type quantum dots

amine functionalized, fluorescence λem 540 nm, 4 μM in 10 mM PBS

Synonym(s):

Fluorescent nanocrystals, QDs

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CdSe/ZnS
UNSPSC Code:
26111700
NACRES:
NA.23

Quality Level

form

dispersion

λmax

525 nm

fluorescence

λem 540 nm
λem 540 nm±10 nm, quantum yield >50%

functional group

amine

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Reactive group: Amine.

Application

The amine functionalized CdS/ZnS or CdSe/ZnS quantum dots (QDs) find applications in bioimaging and biosensing. The three main conjugation approaches reported in literature are conjugation using glutaraldehyde as a linker, conjugations with carboxylic acid containing molecules by using 1-ethyl-3-(3-dimethylaminopropyl) carbodiimide (EDC) coupling, and direct conjugation with isothiocyanates bearing molecules.

Caution

Do not freeze.

Legal Information

Product of Ocean Nanotech, LLC.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kumar P, et al.
Sensors and Actuators B, Chemical, 202, 404-404 (2014)
Jian Wei Tay et al.
Scientific reports, 4, 3918-3918 (2014-01-30)
Phase distortions due to scattering in random media restrict optical focusing beyond one transport mean free path. However, scattering can be compensated for by applying a correction to the illumination wavefront using spatial light modulators. One method of obtaining the
Kavitha Pathakoti et al.
Journal of environmental sciences (China), 25(1), 163-171 (2013-04-17)
Quantum dots (QD) nanoparticles have been widely used in biomedical and electronics fields, because of their novel optical properties. Consequently it confers enormous potential for human exposure and environmental release. To increase the biocompatibility of QDs, a variety of surface
Lily Yang et al.
Small (Weinheim an der Bergstrasse, Germany), 5(2), 235-243 (2008-12-18)
Epidermal growth factor receptor (EGFR) targeted nanoparticle are developed by conjugating a single-chain anti-EGFR antibody (ScFvEGFR) to surface functionalized quantum dots (QDs) or magnetic iron oxide (IO) nanoparticles. The results show that ScFvEGFR can be successfully conjugated to the nanoparticles

Articles

Cadmium-free quantum dots for bioimaging offer design and synthesis strategies, enhancing bioimaging and sensing applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service