Skip to Content
Merck
All Photos(1)

Key Documents

900177

Sigma-Aldrich

Monodispersed nanodiamond particles

10 mg/mL in NMP, carboxylated, 5 nm avg. part. size (DLS)

Synonym(s):

Detonation Nanodiamond, Nanodiamond

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352302

description

-45.1 mV ±20%

Quality Level

form

dispersion
nanoparticles

concentration

10 mg/mL in NMP

avg. part. size

5 nm (DLS)

Looking for similar products? Visit Product Comparison Guide

Application

Detonation nanodiamonds find applications in polymer composites for providing improved mechanical properties, UV protection, and improved resistance to radiation[1]. They also find biological applications extending from drug delivery to positron emission tomography and other imaging techniques[1][2][3][4][5].

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

195.8 °F

Flash Point(C)

91 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The properties and applications of nanodiamonds.
Mochalin VN, et al.
Nature Nanotechnology, 7, 11-11 (2012)
Nanodiamond Particles: Properties and Perspectives for Bioapplications.
Schrand AM, et al.
Critical Reviews in Solid State and Materials Sciences, 34, 18-18 (2009)
Cancer nanomedicine: from drug delivery to imaging.
Chow E, et al.
Science Translational Medicine, 5 (2013)
Functionality is Key: Recent Progress in the Surface Modification of Nanodiamond.
Krueger A and Lang D
Advances in Functional Materials, 22, 890-890 (2012)
The properties and applications of nanodiamonds.
Mochalin VN,et al.
Nature Nanotechnology, 7, 11-11 (2012)

Articles

Among various nanomaterials, nanoparticles of less than 10 nm size remain the most sought-after products.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service