Skip to Content
Merck
All Photos(1)

Key Documents

900163

Sigma-Aldrich

UV-Vis light curable resin

greener alternative

rapid cure, adhesible to plastics and glass

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$243.00

$243.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 G
$243.00

About This Item

UNSPSC Code:
12162002
NACRES:
NA.23

$243.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

form

liquid (colorless)

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

viscosity

360 cP (Brookfield LDTDV-II digital viscometer with size 31 spindle)

transition temp

flash point 110 °F

density

1.17 g/mL

greener alternative category

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Storage:
  • The storage temperature should not exceed 35 °C.
  • Keep tightly closed. Store in a cool, dry, well-ventilated area.
  • Do not expose to ultraviolet light.
  • High temperatures (>80 °C), pressure and contamination with peroxides may result in auto-polymerization of the product.
  • Unstable upon depletion of inhibitor.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product belongs to enabling category of greener alternatives and thus aligns with "Design for energy efficiency". Click here for more information.

Application

  • Cures extremely rapidly to form a glassy, high modulus material.
  • Excellent for use in optical applications.
  • High refractive index - 1.54.
  • High impact absorption.
  • Non-yellowing.
  • No oxygen inhibition.
  • Mechanical Properties after curing:
Modulus*: 100MPa
Strength*: 2.8MPa
Elongation*: 51%
Tack Free**: 130 fpm

*Test resources: Tensile tester, 35mm gauge length, 1.0 mm thickness, 6.5 mm width.
** Cured under stated intensity. Medium pressure mercury bulb with 320-500 nm filter. Adhesion tests were conducted using a cross hatch tape test on films 0.125 mm thick on glass and PET. Material Data using ASTM D 638-02a.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 2

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

230.0 °F

Flash Point(C)

110 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service