Skip to Content
Merck
All Photos(1)

Key Documents

900078

Sigma-Aldrich

Sodium taurodeoxycholate-2,2,4,4,11,11-d6

≥98 atom % D, ≥98% (CP)

Synonym(s):

Sodium deoxytaurocholate-2,2,4,4,11,11-d6, Taurodeoxycholic-2,2,4,4,11,11-d6 acid sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$2,070.00

$2,070.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
5 MG
$2,070.00

About This Item

Linear Formula:
C26D6H38NO6S · Na
Molecular Weight:
527.72
UNSPSC Code:
12352106
NACRES:
NA.12

$2,070.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

isotopic purity

≥98 atom % D

Quality Level

Assay

≥98% (CP)

form

powder

technique(s)

mass spectrometry (MS): suitable

mass shift

M+6

storage temp.

−20°C

SMILES string

C[C@@]12C(CC[C@H]2[C@H](C)CCC(NCCS(=O)(O[Na])=O)=O)C3CCC4C([2H])([2H])[C@H](O)C([2H])([2H])C[C@]4(C)C3C([2H])([2H])[C@@H]1O

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service