Skip to Content
Merck
All Photos(1)

Documents

90000

Sigma-Aldrich

1-(p-Tolyl)ethanol

≥97.0% (GC)

Synonym(s):

4,α-Dimethylbenzyl alcohol, Methyl p-tolyl carbinol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12O
CAS Number:
Molecular Weight:
136.19
Beilstein:
1857562
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.0% (GC)

form

liquid

refractive index

n20/D 1.523

bp

218-220 °C (lit.)

density

0.987 g/mL at 20 °C (lit.)

SMILES string

CC(O)c1ccc(C)cc1

InChI

1S/C9H12O/c1-7-3-5-9(6-4-7)8(2)10/h3-6,8,10H,1-2H3

InChI key

JESIHYIJKKUWIS-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

224.6 °F - closed cup

Flash Point(C)

107 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Siqi Huang et al.
International journal of biological macromolecules, 137, 939-947 (2019-07-08)
The severe preparation process, poor swelling properties and mechanical properties of traditional cellulose and polyvinyl alcohol (PVA) composite hydrogels heavily limited their practical applications. To solve these issues, we use long-chain hydroxyethyl celluloses (HECs) as framework backbones, short-chain PVAs as
Siqi Huang et al.
Carbohydrate polymers, 223, 115080-115080 (2019-08-21)
In this study, hydroxyethyl cellulose (HEC) and polyvinyl alcohol (PVA) as the framework, borax as the cross-linker, and biomass lignin from pulping black liquors and biorefinery as the plasticizer were used to synthesize the lignin-based HEC-PVA (LCP) self-healing conductive hydrogel
Napoleon-Nikolaos Vrettos et al.
Pharmaceutical development and technology, 26(3), 349-361 (2021-01-13)
Hypertension is one of the most common chronic cardiovascular disorders. Sustained-release formulations are developed to maintain drug therapeutic levels throughout the treatment of hypertension, to promote patient compliance and improve patient outcomes. We have developed and tested in in vivo trials
Tahereh Mirmajidi et al.
International journal of pharmaceutics, 596, 120213-120213 (2021-01-26)
Wound healing is a complicated process that takes a long time to complete. The three-layer nanofiber wound dressing containing melatonin is highly expected to show remarkable wound repair by reducing the wound healing time. In this study, chitosan (Cs)-polycaprolactone (PCL)/

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service