Skip to Content
Merck
All Photos(1)

Key Documents

89171

Sigma-Aldrich

6-(Boc-amino)hexyl bromide

≥97.0% (GC)

Synonym(s):

tert-Butyl N-(6-bromohexyl)carbamate

Sign Into View Organizational & Contract Pricing

Select a Size

2.5 G
$670.00

$670.00


Estimated to ship onMay 23, 2025


Request a Bulk Order

Select a Size

Change View
2.5 G
$670.00

About This Item

Empirical Formula (Hill Notation):
C11H22BrNO2
CAS Number:
Molecular Weight:
280.20
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

$670.00


Estimated to ship onMay 23, 2025


Request a Bulk Order

Assay

≥97.0% (GC)

form

liquid

reaction suitability

reagent type: cross-linking reagent

refractive index

n20/D 1.473

density

1.192 g/mL at 20 °C (lit.)

functional group

Boc
amine
bromo

SMILES string

O=C(NCCCCCCBr)OC(C)(C)C

InChI

1S/C11H22BrNO2/c1-11(2,3)15-10(14)13-9-7-5-4-6-8-12/h4-9H2,1-3H3,(H,13,14)

InChI key

NXQXVXILNVTMNA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Building block for preparing analogs of biologically active compounds[1][2]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D. Damour et al.
Bioorganic & Medicinal Chemistry Letters, 6, 1667 -1667 (1996)
T.H. Keller, R. Haener
Helvetica Chimica Acta, 76, 884-884 (1993)
Zaheer Ahmad et al.
Journal of biomaterials science. Polymer edition, 27(1), 40-54 (2015-10-23)
Tumor microenvironment-responsive nano drug delivery vehicles are gaining mounting attention in the field of biomedical sciences. The hypoxic response of the tumorous cells due to very low partial pressure of oxygen (some time less than 2.5 mm of Hg) in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service