Skip to Content
Merck
All Photos(1)

Key Documents

88921

Sigma-Aldrich

Sunflower seed oil from Helianthus annuus

tested according to Ph. Eur.

Synonym(s):

Helianthi annui oleum raffinatum

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21

Agency

USP/NF
tested according to Ph. Eur.

form

oil

application(s)

flavors and fragrances

Looking for similar products? Visit Product Comparison Guide

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Precautionary Statements

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liliana Albertengo et al.
Carbohydrate polymers, 92(2), 1641-1646 (2013-02-13)
Chitosan is a modified, natural carbohydrate polymer derived by deacetylation of chitin. Due to the presence of two functional groups can undergo many chemical modifications. In a previous work we described the synthetic strategy and characterization of a novel soluble
N Tous et al.
Journal of animal science, 90 Suppl 4, 340-342 (2013-02-13)
The aim of this study was to investigate underlying mechanisms of dietary conjugated linoleic acid (CLA) on lipid metabolism in various tissues of pigs. Sixteen gilts (73 ± 3 kg) were fed a control (containing sunflower oil) or an experimental
A Zened et al.
Journal of dairy science, 96(1), 451-459 (2012-11-07)
Trans fatty acids (FA), exhibit different biological properties. Among them, cis-9,trans-11 conjugated linoleic acid has some interesting putative health properties, whereas trans-10,cis-12 conjugated linoleic acid has negative effects on cow milk fat production and would negatively affect human health. In
Nagarajan Lakshmipriya et al.
International journal of food sciences and nutrition, 64(2), 131-139 (2012-10-03)
There is little data on the type of vegetable oil used and the prevalence of metabolic syndrome (MS) in Asian Indians. Food frequency questionnaire was used to document the type of cooking oil in 1875 adults in Chennai city. MS
E Santos-Garcés et al.
Meat science, 93(3), 639-644 (2013-01-01)
X-ray microcomputed tomography (μCT) was used for microstructure analysis on four different types of non-acid pork lean fermented sausages, three of them supplemented with 5% pork back fat, sunflower oil or diacylglycerols (DAGs). The data from the μCT analysis were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service