Skip to Content
Merck
All Photos(1)

Key Documents

88723

Sigma-Aldrich

(R)-1-[(SP)-2-(Dicyclohexylphosphino)ferrocenylethyl]diphenylphosphine

≥97%

Synonym(s):

(1R)-1-(Dicyclohexylphosphino)-2-[(1R)-1-(diphenylphosphino)ethyl]ferrocene (acc to CAS), Josiphos SL-J004-1

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$123.00
1 G
$926.00

$123.00


Only 0 left in stock

Request a Bulk Order

Select a Size

Change View
100 MG
$123.00
1 G
$926.00

About This Item

Empirical Formula (Hill Notation):
C36H44FeP2
CAS Number:
Molecular Weight:
594.53
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$123.00


Only 0 left in stock

Request a Bulk Order

Quality Level

Assay

≥97%

form

crystals

optical purity

ee: ≥99%

functional group

phosphine

SMILES string

[Fe].[CH]1[CH][CH][CH][CH]1.C[C@H]([C]2[CH][CH][CH][C]2P(C3CCCCC3)C4CCCCC4)P(c5ccccc5)c6ccccc6

InChI

1S/C31H39P2.C5H5.Fe/c1-25(32(26-15-6-2-7-16-26)27-17-8-3-9-18-27)30-23-14-24-31(30)33(28-19-10-4-11-20-28)29-21-12-5-13-22-29;1-2-4-5-3-1;/h2-3,6-9,14-18,23-25,28-29H,4-5,10-13,19-22H2,1H3;1-5H;/t25-;;/m1../s1

InChI key

AWOOIDCKLIQAJV-KHZPMNTOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

sold in collaboration with Solvias AG

Application

(R)-1-[(SP)-2-(Dicyclohexylphosphino)ferrocenylethyl]diphenylphosphine can be used as a ligand:
  • To prepare iridium metal complex (IrIJOSIPHOS), which is used as a catalyst in the asymmetric hydrogenation of 2-phenylquinoxaline.[1]
  • In the preparation of natural products by asymmetric conjugate addition of Grignard reagents to α-substituted cyclic alkenones.[2]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Catalytic enantioselective conjugate addition en route to paxilline indoloterpenoids
Schatz DJ, et al.
Tetrahedron, 75(24), 3361-3365 (2019)
Additive effects of amines on asymmetric hydrogenation of quinoxalines catalyzed by chiral iridium complexes
Nagano T, et al.
Chemistry?A European Journal , 18(37), 11578-11592 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service