Skip to Content
Merck
All Photos(1)

Documents

88670

Sigma-Aldrich

2-Ethylhexyl thioglycolate

≥95.0% (GC)

Synonym(s):

2-EHTG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2COOCH2CH(C2H5)(CH2)3CH3
CAS Number:
Molecular Weight:
204.33
Beilstein:
9047474
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0% (GC)

form

liquid

refractive index

n20/D 1.461

bp

255-260 °C (lit.)

density

0.972 g/mL at 20 °C (lit.)

SMILES string

CCCCC(CC)COC(=O)CS

InChI

1S/C10H20O2S/c1-3-5-6-9(4-2)7-12-10(11)8-13/h9,13H,3-8H2,1-2H3

InChI key

OWHSTLLOZWTNTQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Ethylhexyl thioglycolate can be used:
  • In the selective extraction of indium(III) and gallium(III).
  • As a reagent in the synthesis of conjugated polymers based on thienoquinoidal building block.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

243.5 °F - Pensky-Martens closed cup

Flash Point(C)

117.5 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel π -Conjugated Polymer Based on an Extended Thienoquinoid
Cai Y, et al.
Chemistry of Materials, 30(2), 319-323 (2018)
Selective extraction of indium (iii) and gallium (iii) with 2-ethylhexyl thioglycolate
Miura R, et al.
Solvent Extraction Research and Development, Japan, 24(2), 123-130 (2017)
Neepa Choksi et al.
Cutaneous and ocular toxicology, 39(3), 180-192 (2020-06-27)
OptiSafe is an in chemico test method that identifies potential eye irritants based on macromolecular damage following test chemical exposure. The OptiSafe protocol includes a prescreen assessment that identifies test chemicals that are outside the applicability domain of the test

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service