Skip to Content
Merck
All Photos(1)

Key Documents

88310

Sigma-Aldrich

Theophylline-7-acetic acid

≥99.0% (T)

Synonym(s):

1,3-Dimethylxanthine-7-acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10N4O4
CAS Number:
Molecular Weight:
238.20
Beilstein:
279221
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0% (T)

form

powder

SMILES string

CN1C(=O)N(C)c2ncn(CC(O)=O)c2C1=O

InChI

1S/C9H10N4O4/c1-11-7-6(8(16)12(2)9(11)17)13(4-10-7)3-5(14)15/h4H,3H2,1-2H3,(H,14,15)

InChI key

HCYFGRCYSCXKNQ-UHFFFAOYSA-N

Gene Information

human ... ADORA2B(136)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Grosa et al.
Drug metabolism and disposition: the biological fate of chemicals, 14(2), 267-270 (1986-03-01)
The metabolic transformation of the bronchospasmolytic agent doxophylline (2-(7'-theophyllinemethyl)-1,3-dioxolane) was studied in vitro with phenobarbital-induced rat liver microsomal fraction containing the NADPH-generating system. Doxophylline was poorly metabolized as 95% of the recovered material was parent compound. The major metabolite resulted:
C Ferretti et al.
International journal of tissue reactions, 14(1), 31-36 (1992-01-01)
It is assumed that theophylline (THEO) and its xanthinic derivatives inhibit lung phosphodiesterase (PDE) and block adenosine receptors in the induction of bronchodilatation. Since the theophyllinic compound ambroxol-theophylline-7-acetic acid (ATA) has been shown in vivo to be a sound bronchodilator
J G Ufkes et al.
Archives internationales de pharmacodynamie et de therapie, 253(2), 301-314 (1981-10-01)
A study has been made of the bronchospasmolytic actions of theophylline and some ot its N-7-substituted derivatives administered by i.v infusion in anaesthetized guinea pigs, in which experimental bronchial asthma was induced by i.v. administration of histamine, 5-hydroxytryptamine and bradykinin.
G Dănilă et al.
Revista medico-chirurgicala a Societatii de Medici si Naturalisti din Iasi, 94(3-4), 581-586 (1990-10-01)
Some esters of 7-theophyllinylacetic acid were synthetized, characterized physicochemically and tested for their anti-inflammatory properties. As compared to indomethacin, reference anti-inflammatory drug, all synthetized compounds were less toxic. The anti-inflammatory properties are influenced by the nature of the ester group
Gas-liquid chromatographic determination of acephylline in urine.
J Zuidema et al.
Journal of chromatography, 182(3-4), 445-447 (1980-06-13)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service