Skip to Content
Merck
All Photos(1)

Key Documents

861847

Sigma-Aldrich

Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone

97%

Synonym(s):

FCCP, Mesoxalonitrile 4-trifluoromethoxyphenylhydrazone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3OC6H4NHN=C(CN)2
CAS Number:
Molecular Weight:
254.17
Beilstein:
664446
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

97%

mp

174-175 °C (dec.) (lit.)

SMILES string

FC(F)(F)Oc1ccc(N\N=C(/C#N)C#N)cc1

InChI

1S/C10H5F3N4O/c11-10(12,13)18-9-3-1-7(2-4-9)16-17-8(5-14)6-15/h1-4,16H

InChI key

BMZRVOVNUMQTIN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lin Du et al.
Journal of natural products, 73(11), 1868-1872 (2010-10-12)
The mammea-type coumarin mammea E/BB (1) was found to inhibit both hypoxia-induced and iron chelator-induced hypoxia-inducible factor-1 (HIF-1) activation in human breast tumor T47D cells with IC(50) values of 0.96 and 0.89 μM, respectively. Compound 1 suppressed the hypoxic induction
A Magge et al.
Journal of applied microbiology, 106(3), 814-824 (2009-02-04)
To determine roles of coats in staining Bacillus subtilis spores, and whether spores have membrane potential. Staining by four dyes and autofluorescence of B. subtilis spores that lack some (cotE, gerE) or most (cotE gerE) coat protein was measured. Wild-type
Robin K Minor et al.
Scientific reports, 1, 70-70 (2012-02-23)
Sirt1 is an NAD(+)-dependent deacetylase that extends lifespan in lower organisms and improves metabolism and delays the onset of age-related diseases in mammals. Here we show that SRT1720, a synthetic compound that was identified for its ability to activate Sirt1
Dawn A Delfín et al.
Bioorganic & medicinal chemistry, 17(2), 820-829 (2008-12-09)
BTB 06237 (2-[(2,4-dichloro-5-methylphenyl)sulfanyl]-1,3-dinitro-5-(trifluoromethyl) benzene), a compound previously identified through QSAR pharmacophore development and a virtual screen of the Maybridge database, possesses potent and selective activity against Leishmania parasites. In the present study, several analogs of BTB 06237 were synthesized and
Sean Ekins et al.
Drug metabolism and disposition: the biological fate of chemicals, 38(12), 2302-2308 (2010-09-17)
Drug-induced liver injury (DILI) is one of the most important reasons for drug development failure at both preapproval and postapproval stages. There has been increased interest in developing predictive in vivo, in vitro, and in silico models to identify compounds

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service