Skip to Content
Merck
All Photos(1)

Key Documents

860794

Sigma-Aldrich

Z-L-Phe chloromethyl ketone

98%

Synonym(s):

N-Carbobenzyloxy-L-phenylalanyl chloromethyl ketone, NSC 251810, SL-01, ZPCK

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$339.00

$339.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$339.00

About This Item

Linear Formula:
C6H5CH2CH(NHCO2CH2C6H5)COCH2Cl
CAS Number:
Molecular Weight:
331.79
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.22

$339.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

powder

optical activity

[α]23/D +30°, c = 1 in chloroform

reaction suitability

reaction type: solution phase peptide synthesis

mp

107-108 °C (lit.)

application(s)

peptide synthesis

storage temp.

−20°C

SMILES string

ClCC(=O)[C@H](Cc1ccccc1)NC(=O)OCc2ccccc2

InChI

1S/C18H18ClNO3/c19-12-17(21)16(11-14-7-3-1-4-8-14)20-18(22)23-13-15-9-5-2-6-10-15/h1-10,16H,11-13H2,(H,20,22)/t16-/m0/s1

InChI key

OYHLRJGDELITAF-INIZCTEOSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Enzyme inhibitor.[1]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Jung et al.
Protein science : a publication of the Protein Society, 4(11), 2433-2435 (1995-11-01)
The essential histidine residue of carboxypeptidase Y (CPY) was modified by a site-specific reagent, a chloromethylketone derivative of benzyloxycarbonyl-L-phenylalanine. The single modified histidine residue was converted to N tau-carboxy-methyl histidine (cmHis) upon performic acid oxidation. A peptide containing cmHis was
L A Johnson et al.
Biochimica et biophysica acta, 953(3), 269-279 (1988-04-14)
The reaction of chymase, a chymotryptic proteinase from human skin, and bovine pancreatic chymotrypsin with a number of time-dependent inhibitors has been studied. An integrated equation, relating product formation with time, has been derived for the reaction of enzymes with
Inhibition of carboxypeptidase Y by chloromethyl ketone derivatives of benzyloxycarbonyl-L-phenylalanine.
R Hayashi et al.
Journal of biochemistry, 76(6), 1355-1357 (1974-12-01)
Biogenesis of vaccina: interrelationship between post-translational cleavage, virus assembly, and maturation.
M Silver et al.
Virology, 117(2), 341-356 (1982-03-01)
T Wileman et al.
Cell regulation, 2(9), 753-765 (1991-09-01)
The endoplasmic reticulum, or an organelle closely associated with it, contains proteases that can be used to remove partially assembled or improperly folded proteins. Very little is known at present about the types of protease that degrade these proteins. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service