Skip to Content
Merck
All Photos(1)

Key Documents

860263

Sigma-Aldrich

Methyl α-D-mannopyranoside

99%

Synonym(s):

α-Methyl D-mannoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14O6
CAS Number:
Molecular Weight:
194.18
Beilstein:
81566
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

99%

optical activity

[α]22/D +79.4°, c = 1 in H2O

mp

193-196 °C (lit.)

SMILES string

CO[C@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@@H]1O

InChI

1S/C7H14O6/c1-12-7-6(11)5(10)4(9)3(2-8)13-7/h3-11H,2H2,1H3/t3-,4-,5+,6+,7+/m1/s1

InChI key

HOVAGTYPODGVJG-VEIUFWFVSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used to reduce Con A binding to lymphocyte surface in culture.[1]

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E C Snow et al.
Journal of immunology (Baltimore, Md. : 1950), 124(2), 739-744 (1980-02-01)
The ability of insulin to influence the responsiveness of murine T lymphocytes in a culture system containing a serum substitute was documented. The presence of insulin was found to enhance the concanavalin A (Con A) reactivity of the lymphocytes. Once

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service