Skip to Content
Merck
All Photos(1)

Key Documents

859370

Sigma-Aldrich

DL-5-Hydroxylysine hydrochloride

95%

Synonym(s):

2,6-Diamino-5-hydroxycaproic acid hydrochloride, 2,6-Diamino-5-hydroxyhexanoic acid hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CH(OH)CH2CH2CH(NH2)COOH · HCl
CAS Number:
Molecular Weight:
198.65
Beilstein:
3914368
EC Number:
MDL number:
UNSPSC Code:
12352200

Assay

95%

mp

225 °C (dec.) (lit.)

SMILES string

Cl.NCC(O)CCC(N)C(O)=O

InChI

1S/C6H14N2O3.ClH/c7-3-4(9)1-2-5(8)6(10)11;/h4-5,9H,1-3,7-8H2,(H,10,11);1H

InChI key

MJXVOTKVFFAZQJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

A constituent of collagen.

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service