850926
2′,3′,5′-Tri-O-acetylguanosine
98%
Synonym(s):
Guanosine 2′,3′,5′-triacetate
About This Item
Recommended Products
Assay
98%
form
powder
mp
226-231 °C (lit.)
storage temp.
2-8°C
SMILES string
CC(=O)OC[C@H]1O[C@H]([C@H](OC(C)=O)[C@@H]1OC(C)=O)n2cnc3C(=O)NC(N)=Nc23
InChI
1S/C16H19N5O8/c1-6(22)26-4-9-11(27-7(2)23)12(28-8(3)24)15(29-9)21-5-18-10-13(21)19-16(17)20-14(10)25/h5,9,11-12,15H,4H2,1-3H3,(H3,17,19,20,25)/t9-,11-,12-,15-/m1/s1
InChI key
ULXDFYDZZFYGIY-SDBHATRESA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service