Skip to Content
Merck
All Photos(1)

Key Documents

850276

Sigma-Aldrich

4-Fluorophenoxyacetic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4OCH2CO2H
CAS Number:
Molecular Weight:
170.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

104-104.5 °C (lit.)

SMILES string

OC(=O)COc1ccc(F)cc1

InChI

1S/C8H7FO3/c9-6-1-3-7(4-2-6)12-5-8(10)11/h1-4H,5H2,(H,10,11)

InChI key

ZBIULCVFFJJYTN-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chayanis Sutcharitchan et al.
Food chemistry, 322, 126766-126766 (2020-04-20)
A simple, rapid, and effective method based on salting-out extraction and LC-MS/MS techniques was developed for the determination of 39 plant growth regulator (PGR) residues in two of the most common root and rhizome Chinese herbs, Codonopsis Radix (Dangshen) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service