Skip to Content
Merck
All Photos(1)

Key Documents

83086

Sigma-Aldrich

1-Butyl-3-methylimidazolium methyl sulfate

≥97.0% (HPLC)

Synonym(s):

BMIMMeSO4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H18N2O4S
CAS Number:
Molecular Weight:
250.32
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (HPLC)

form

liquid

refractive index

n20/D 1.476-1.482

density

1.21 g/mL at 20 °C

SMILES string

COS([O-])(=O)=O.CCCCn1cc[n+](C)c1

InChI

1S/C8H15N2.CH4O4S/c1-3-4-5-10-7-6-9(2)8-10;1-5-6(2,3)4/h6-8H,3-5H2,1-2H3;1H3,(H,2,3,4)/q+1;/p-1

InChI key

MEMNKNZDROKJHP-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Other Notes

Halogen free ionic liquid[1][2]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T. Itoh et al.
Chemistry Letters (Jpn), 32, 654-654 (2003)
J.S. Yadav et al.
New. J. Chem., 27, 462-462 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service