Skip to Content
Merck
All Photos(1)

Key Documents

81391

Sigma-Aldrich

Poly(4-vinylpyridine), cross-linked

2 % cross-linked with divinylbenzene

Synonym(s):

Reillex® 402

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$132.00
100 G
$332.00

$132.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$132.00
100 G
$332.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

$132.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

crosslinking

2 % cross-linked with divinylbenzene

loss

≤10% loss on drying, 110°C

SMILES string

n2ccc(cc2)C=C.c1(c(cccc1)C=C)C=C

InChI

1S/C10H10.C7H7N/c1-3-9-7-5-6-8-10(9)4-2;1-2-7-3-5-8-6-4-7/h3-8H,1-2H2;2-6H,1H2

InChI key

HUEZUDXJCGHIHG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Reillex is a registered trademark of Vertellus Specialties, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service