Skip to Content
Merck
All Photos(1)

Key Documents

809756

Sigma-Aldrich

Vitamin D3-25,26,27-13C3 solution

100 μg/mL in ethanol, ≥98 atom % 13C, ≥97% (CP)

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$1,210.00

$1,210.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request more information

Select a Size

Change View
1 ML
$1,210.00

About This Item

Empirical Formula (Hill Notation):
13C3C24H44O
Molecular Weight:
387.62
EC Number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.12

$1,210.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request more information

isotopic purity

≥98 atom % 13C

Quality Level

Assay

≥97% (CP)

form

solution

concentration

100 μg/mL in ethanol

technique(s)

mass spectrometry (MS): suitable

color

colorless

mass shift

M+3

storage temp.

−20°C

SMILES string

C=C1CC[C@H](O)C/C1=C/C=C2C(CC[C@@H]3[C@@H](CCC[13CH]([13CH3])[13CH3])C)[C@]3(C)CCC/2

InChI

1S/C27H44O/c1-19(2)8-6-9-21(4)25-15-16-26-22(10-7-17-27(25,26)5)12-13-23-18-24(28)14-11-20(23)3/h12-13,19,21,24-26,28H,3,6-11,14-18H2,1-2,4-5H3/b22-12+,23-13-/t21-,24+,25-,26?,27-/m1/s1/i1+1,2+1,19+1

InChI key

QYSXJUFSXHHAJI-ZGZAZNHXSA-N

Related Categories

General description

Vitamin D3, also known as cholecalciferol, promotes the absorption of calcium and phosphorus in the body to regulate bone growth. Vitamin D3 is most commonly found in humans and animals and is produced in our skin when exposed to sunlight. It is used to treat and prevent bone disorders (such as rickets, osteomalacia).[1][2]
Vitamin D3-25,26,27-13Cis an isotope of vitamin D3 wherein C-25, C-26, C-27 carbons are replaced by 13C6 isotope.

Application

Vitamin D3-25,26,27-13C3 can be used as an analytical standard for accurate data interpretation in specific biological samples.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F

Flash Point(C)

14 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vitamin D is not directly necessary for bone growth and mineralization
Underwood JL and DeLuca HF
American Journal of Physiology. Endocrinology and Metabolism, 246(6), E493-E498 (1984)
Effect of four monthly oral vitamin D3 (cholecalciferol) supplementation on fractures and mortality in men and women living in the community: randomised double blind controlled trial
Trivedi DP, et al.
BMJ (Clinical Research ed.), 326(7387), 469-469 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service