Skip to Content
Merck
All Photos(2)

Key Documents

809551

Sigma-Aldrich

Methyltriphenylphosphonium methylcarbonate

Synonym(s):

Perosa-Selva-Noè Vinylation Reagent

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$379.00

$379.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$379.00

About This Item

Empirical Formula (Hill Notation):
C21H21O3P
CAS Number:
Molecular Weight:
352.36
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

$379.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

reaction suitability

reaction type: C-C Bond Formation

mp

146-151 °C

functional group

phosphine

storage temp.

2-8°C

SMILES string

C[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.[O-]C(OC)=O

InChI

1S/C19H18P.C2H4O3/c1-20(17-11-5-2-6-12-17,18-13-7-3-8-14-18)19-15-9-4-10-16-19;1-5-2(3)4/h2-16H,1H3;1H3,(H,3,4)/q+1;/p-1

InChI key

FALLFMKOBMQQNQ-UHFFFAOYSA-M

General description

Methyltriphenylphosphonium methylcarbonate, also known as Perosa-Selva-Noè vinylation reagent, is a green reagent for performing halide and base-free Wittig reaction. It can be prepared by reacting triphenylphosphine with dimethylcarbonate.[1]

Application

[Ph3PCH3][CH3OCO2] is a latent ylide that promotes Wittig vinylation of aldehydes and ketones. Alkenes are obtained simply by mixing [Ph3PCH3][CH3OCO2] and the carbonyl and heating in a solvent (no base, no halides, and no inorganic byproducts needed).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Methyltriphenylphosphonium Methylcarbonate, an All-In-One Wittig Vinylation Reagent.
Cattelan L, et al.
ChemSusChem, 8(23), 3963-3966 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service