Skip to Content
Merck
All Photos(5)

Key Documents

806633

Sigma-Aldrich

Graphene nanoplatelets

powder, hydrocarbon functionalized, hydrophobic

Synonym(s):

GNPs, graphenit-LIPO

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
UNSPSC Code:
12352103
NACRES:
NA.23

description

Dispersibility: dichloromethane, N-methyl-2-pyrrolidone, and other non-polar solvents

form

powder

composition

Carbon, >85 wt. %
Oxygen, >3 wt. %

Looking for similar products? Visit Product Comparison Guide

General description

Graphene nanoplatelets are “pure carbon”, with a high tendency to aggregate and poor interactions with the surrounding matrix, hence difficult to disperse in processes and supports such as epoxy resin, or thermoplastics. The chemical modification on the surface of graphite nanoplatelet open new properties allowing good interaction with the targeted matrix. Without these chemical modifications the dispersion step became a difficult task and quite often do not produce the expected benefits.

Application

  • Graphene (nano)composite materials
  • Conductive coatings
  • Conductive Inks
  • Energy Storage
  • Electrode materials
  • Room Temperature

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Graphene Modified Lipophilically by Stearic Acid and its Composite With Low Density Polyethylene
Han et al.
Journal of Macromolecular Science: Physics, Part B: Physics, 53(7), 1193-1204 (2014)
Sheng-Zhen Zu et al.
Journal of nanoscience and nanotechnology, 13(2), 946-953 (2013-05-08)
Graphene sheets have been dispersed in water and organic solvents by noncovalent surface modification with the assistance of bifunctional molecule pyrene-adamantane (Py-Ad) carrying a pyrene moiety. Lipophilic graphene sheets have been obtained in the form of graphene-Py-Ad in a wide

Articles

3D printing is a type of additive manufacturing that can be used to rapidly fabricate components with highly customizable geometries.

Since its discovery little more than a decade ago,1 the two-dimensional (2D) allotrope of carbon—graphene—has been the subject of intense multidisciplinary research efforts.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service