Skip to Content
Merck
All Photos(1)

Documents

806609

Sigma-Aldrich

Silver

nanowires, diam. × L 20 nm (±2 nm) × 12 μm (±2 μm), 5 mg/mL (in water)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12141740
NACRES:
NA.23

description

Flash point >230 °F

Quality Level

form

nanowires
suspension

concentration

5 mg/mL (in water)

diam. × L

20 nm (±2 nm) × 12 μm (±2 μm)

color

gray

density

0.999 g/mL

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Owing to unique physical and chemical properties the nanowires of silver find wide range of applications extending from catalysis to photovoltaics, touch screen display, biosensing etc.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dzmitry Afanasenkau et al.
Nature biomedical engineering, 4(10), 1010-1022 (2020-09-23)
Neuromuscular interfaces are required to translate bioelectronic technologies for application in clinical medicine. Here, by leveraging the robotically controlled ink-jet deposition of low-viscosity conductive inks, extrusion of insulating silicone pastes and in situ activation of electrode surfaces via cold-air plasma

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service