Skip to Content
Merck
All Photos(4)

Key Documents

806196

Sigma-Aldrich

Benzylammonium iodide

greener alternative

Synonym(s):

Greatcell Solar®, Phenylmethylamine iodide

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$905.00

$905.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$905.00

About This Item

Empirical Formula (Hill Notation):
C7H10IN
Molecular Weight:
235.07
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.23

$905.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

Quality Level

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

179.35 °C

greener alternative category

SMILES string

NCC1=CC=CC=C1.I

InChI

1S/C7H9N.HI/c8-6-7-4-2-1-3-5-7;/h1-5H,6,8H2;1H

InChI key

PPCHYMCMRUGLHR-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more details.

Application

Benzylammonium iodide can be used as an additive to improve the crystal formation of perovskites, which further facilitate an increase in the efficiency of solar cells.[1][2] It can also be used in the formation of alkaline exchange membrane based fuel cells.[3]
The iodide and bromide based alkylated halides find applications as precursors for fabrication of perovskites for photovoltaic applications.[4][5][6]

Legal Information

Product of Greatcell Solar®
Greatcell Solar is a registered trademark of Greatcell Solar

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mechanistic analysis of ammonium cation stability for alkaline exchange membrane fuel cells
Mohanty AD and Bae C
Journal of Material Chemistry A, 2(41), 17314-17320 (2014)
Ammonium-iodide-salt additives induced photovoltaic performance enhancement in one-step solution process for perovskite solar cells
Yang Y, et al.
Journal of alloys and compounds, 684, 84-90 (2016)
Efficient solar cells with enhanced humidity and heat stability based on benzylammonium-caesium-formamidinium mixed-dimensional perovskites
Liu G, et al.
Journal of Material Chemistry A, 6(37), 18067-18074 (2018)
So-Yeon Kim et al.
Nanoscale, 11(30), 14330-14338 (2019-07-20)
We report here the effect of interlayer spacing in 2-dimensional (2D) perovskites of [C6H5(CH2)nNH3]2PbI4 (anilinium (An) for n = 0, benzylammonium (BzA) for n = 1 and phenylethylammonium (PEA) for n = 2) on resistive switching properties. X-ray diffraction (XRD)
Nam Joong Jeon et al.
Nature, 517(7535), 476-480 (2015-01-07)
Of the many materials and methodologies aimed at producing low-cost, efficient photovoltaic cells, inorganic-organic lead halide perovskite materials appear particularly promising for next-generation solar devices owing to their high power conversion efficiency. The highest efficiencies reported for perovskite solar cells

Articles

Next generation solar cells have the potential to achieve conversion efficiencies beyond the Shockley-Queisser (S-Q) limit while also significantly lowering production costs.

Dr. Perini and Professor Correa-Baena discuss the latest research and effort to obtain higher performance and stability of perovskite materials.

For several decades, the need for an environmentally sustainable and commercially viable source of energy has driven extensive research aimed at achieving high efficiency power generation systems that can be manufactured at low cost.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service