Skip to Content
Merck
All Photos(2)

Key Documents

804746

Sigma-Aldrich

Poly(N-(2-hydroxypropyl)methacrylamide)

average Mn 30,000-50,000, PDI ≤1.3

Synonym(s):

PHPMA, PolyHPMA

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$272.00

$272.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$272.00

About This Item

Linear Formula:
(C7H13NO2)n
UNSPSC Code:
12162002
NACRES:
NA.23

$272.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

Quality Level

mol wt

average Mn 30,000-50,000

PDI

≤1.3

storage temp.

2-8°C

Application

PolyHPMA is a biocompatible, hydrophilic polymer commonly used in bioconjugation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jindrich Kopecek et al.
Advanced drug delivery reviews, 62(2), 122-149 (2009-11-19)
The overview covers the discovery of N-(2-hydroxypropyl)methacrylamide (HPMA) copolymers, initial studies on their synthesis, evaluation of biological properties, and explorations of their potential as carriers of biologically active compounds in general and anticancer drugs in particular. The focus is on

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service