Skip to Content
Merck
All Photos(1)

Key Documents

804681

Sigma-Aldrich

Potassium 5-hydroxypentanoyltrifluoroborate

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$179.00

$179.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$179.00

About This Item

Empirical Formula (Hill Notation):
C5H9BF3KO2
CAS Number:
Molecular Weight:
208.03
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$179.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

Quality Level

mp

123.6 °C

functional group

hydroxyl

SMILES string

[K+].OCCCCC(=O)[B-](F)(F)F

InChI

1S/C5H9BF3O2.K/c7-6(8,9)5(11)3-1-2-4-10;/h10H,1-4H2;/q-1;+1

InChI key

OLLDPQQFLCSTQN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Potassium 5-hydroxypentanoyltrifluoroborate belongs to the class of compounds known as potassium acyltrifluroborates (KATs). It acts as an acyl donor during the amidation of O-benzoyl hydroxylamine to form the corresponding amide product.[1]

Application

Potassium Acyltrifluroborates (KATs) are bench, air and moisture stable reagents for rapid, chemoselective amide bond formations with hydroxylamines. These amide bond forming reactions proceed under aqueous conditions, without the need for coupling reagents or protecting groups. This product was introduced in collaboration with the Bode Research Group.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amide-forming ligation of acyltrifluoroborates and hydroxylamines in water.
Aaron M Dumas et al.
Angewandte Chemie (International ed. in English), 51(23), 5683-5686 (2012-04-28)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service