803987
N-(4-(benzyloxy)-2-methylbutan-2-yl)-2-chloroaniline
About This Item
form
liquid
storage temp.
2-8°C
SMILES string
CC(CCOCC1=CC=CC=C1)(C)NC2=C(Cl)C=CC=C2
InChI
1S/C18H22ClNO/c1-18(2,20-17-11-7-6-10-16(17)19)12-13-21-14-15-8-4-3-5-9-15/h3-11,20H,12-14H2,1-2H3
InChI key
KEXYAYZJBMLTHS-UHFFFAOYSA-N
Application
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service