Skip to Content
Merck
All Photos(1)

Documents

803928

Sigma-Aldrich

Benzyl 4-(trifluoromethylsulfonyloxy)-3-(trimethylsilyl)-5,6-dihydropyridine-1(2H)-carboxylate

Synonym(s):

Garg Azacyclohexyne Precursor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H22F3NO5SSi
Molecular Weight:
437.51
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

description

FP: >230°F

form

liquid

refractive index

n20/D 1.4902

density

1.2625 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

C[Si](C)(C)C1=C(OS(=O)(C(F)(F)F)=O)CCN(C(OCC2=CC=CC=C2)=O)C1

InChI

1S/C17H22F3NO5SSi/c1-28(2,3)15-11-21(16(22)25-12-13-7-5-4-6-8-13)10-9-14(15)26-27(23,24)17(18,19)20/h4-8H,9-12H2,1-3H3

InChI key

QGKRRFLQHRMSAL-UHFFFAOYSA-N

General description

Benzyl 4-(trifluoromethylsulfonyloxy)-3-(trimethylsilyl)-5,6-dihydropyridine-1(2H)-carboxylate is a silyl triflate. It can be prepared from 4-methoxypyridine, via a three-step route.

Application

Benzyl 4-(trifluoromethylsulfonyloxy)-3-(trimethylsilyl)-5,6-dihydropyridine-1(2H)-carboxylate may be used as a starting reagent for the synthesis of 3,4-piperidyne. 3,4-Piperidyne is a precursor for the synthesis of various functionalized heterocyclic compounds.
Building block is a precursor for a highly useful azacyclohexyne intermediate, which is generated in the presence of Cesium Fluoride. The azacyclohexyne can be trapped in situ with dipolariphiles and other dienes to form bicyclic carbocycles and heterocycles.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Travis C McMahon et al.
Journal of the American Chemical Society, 137(12), 4082-4085 (2015-03-15)
We report the generation of the first 3,4-piperidyne and its use as a building block for the synthesis of annulated piperidines. Experimental and computational studies of this intermediate are disclosed, along with comparisons to the well-known 3,4-pyridyne. The distortion/interaction model

Related Content

The Garg group develops methods for the synthesis of natural products and pharmaceuticals. One key method pertains to heterocyclic arynes, such as indolynes and pyridynes, which are generated in situ from silyltriflate precursors under mild fluoride based reaction conditions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service