Skip to Content
Merck
All Photos(1)

Key Documents

802921

Sigma-Aldrich

Estrone-2,3,4-13C3 solution

100 μg/mL in methanol, 99 atom % 13C, 98% (CP)

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$293.00

$293.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
1 ML
$293.00

About This Item

Empirical Formula (Hill Notation):
13C3C15H22O2
CAS Number:
Molecular Weight:
273.34
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.12

$293.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

isotopic purity

99 atom % 13C

Quality Level

Assay

98% (CP)

form

solution

concentration

100 μg/mL in methanol

technique(s)

mass spectrometry (MS): suitable

mass shift

M+3

shipped in

dry ice

SMILES string

C[C@@]12C(C[C@@H](O)[C@@H]2O)C3CCC4=[13CH][13C](O)=[13CH]C=C4C3CC1

InChI

1S/C18H24O3/c1-18-7-6-13-12-5-3-11(19)8-10(12)2-4-14(13)15(18)9-16(20)17(18)21/h3,5,8,13-17,19-21H,2,4,6-7,9H2,1H3/t13?,14?,15?,16-,17+,18+/m1/s1/i3+1,8+1,11+1

InChI key

PROQIPRRNZUXQM-USIGXHLHSA-N

Related Categories

Application

This product may be used as an analytical standard.

Packaging

1 mL in ampule
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service