Skip to Content
Merck
All Photos(1)

Key Documents

80137

Sigma-Aldrich

Diisopropyl phthalate

≥97.0% (GC)

Synonym(s):

1,2-Benzenedicarboxylic acid 1,2-bis(1-methylethyl) ester, NSC 15315, Phthalic acid diisopropyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
$561.00

$561.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 ML
$561.00

About This Item

Empirical Formula (Hill Notation):
C14H18O4
CAS Number:
Molecular Weight:
250.29
Beilstein:
1972723
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$561.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥97.0% (GC)

form

solid

refractive index

n20/D 1.490 (lit.)

acid value

≤2

density

1.063 g/mL at 20 °C (lit.)

functional group

ester

SMILES string

CC(C)OC(=O)c1ccccc1C(=O)OC(C)C

InChI

1S/C14H18O4/c1-9(2)17-13(15)11-7-5-6-8-12(11)14(16)18-10(3)4/h5-10H,1-4H3

InChI key

QWDBCIAVABMJPP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei Jia et al.
Journal of chromatography. A, 1362, 110-118 (2014-08-27)
A new analytical method was developed and validated for simultaneous analysis of 27 phthalates in milk and milk products. Response surface methodology was employed to optimize a quick, easy, cheap, effective, rugged, and safe (QuEChERS) sample preparation method. Ultrahigh-performance liquid

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service