Skip to Content
Merck
All Photos(1)

Key Documents

799637

Sigma-Aldrich

2-n-Butyl-2-oxazoline

Synonym(s):

2-Butyl-2-oxazoline, 2-Butyl-4,5-dihydrooxazole

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$204.00

$204.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$204.00

About This Item

Empirical Formula (Hill Notation):
C7H13NO
CAS Number:
Molecular Weight:
127.18
Beilstein:
107201
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$204.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

liquid

Quality Level

storage temp.

2-8°C

SMILES string

CCCCC1=NCCO1

InChI

1S/C7H13NO/c1-2-3-4-7-8-5-6-9-7/h2-6H2,1H3

InChI key

VERUITIRUQLVOC-UHFFFAOYSA-N

General description

2-n-Butyl-2-oxazoline is a member of the 2-oxazoline family used in the synthesis of well-defined polymers with controlled molecular weights and narrow molecular weight distributions. It is used particularly through its application in cationic ring-opening polymerization to produce poly(2-oxazoline)s. Its unique properties facilitate advancements in biomedical applications, functional materials development.

Application

2-n-Butyl-2-oxazoline can be used as:
  • A monomer in the synthesis of poly(2-oxazoline)s, which are promising candidates for drug delivery systems, tissue engineering scaffolds, and other biomedical uses. Their ability to form hydrophilic or amphiphilic structures enhances drug encapsulation and release profiles.
  • A monomer in the synthesis of core cross-linked star (CCS) polymers, where it serves as a hydrophobic component. These CCS polymers exhibit improved drug loading capacities and release behaviors, making them suitable for targeted therapy applications.
  • A key monomer in the synthesis in the design and synthesis of eco-friendly colorimetric nanofibers. These nanofibrous sensors are used for colorimetric pH detection.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

73.4 °F

Flash Point(C)

23 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

PiPrOx-based polymers exhibit diverse stimuli-responsive properties, showcased in recent developments for functional polymer systems.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service