Skip to Content
Merck
All Photos(1)

Key Documents

799173

Sigma-Aldrich

Dimethyl-di(methacroyloxy-1-ethoxy)silane

>90.0%

Synonym(s):

2-Propenoic acid, 2-methyl-, 1,1′-[(dimethylsilylene)bis(oxy-2,1-ethanediyl)] ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H24O6Si
CAS Number:
Molecular Weight:
316.42
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

>90.0%

form

liquid

contains

50-500 ppm Phenothiazine as inhibitor

density

1.046 at 25 °C

storage temp.

2-8°C

SMILES string

CC(C(OCCO[Si](C)(C)OCCOC(C(C)=C)=O)=O)=C

InChI

1S/C14H24O6Si/c1-11(2)13(15)17-7-9-19-21(5,6)20-10-8-18-14(16)12(3)4/h1,3,7-10H2,2,4-6H3

InChI key

OWETURSZOBVTAD-UHFFFAOYSA-N

Application

UV curable silane based cross-linkers for preparation of methyl methacrylate hydrogels for medical devices.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and Characterization of Star Polymers and Cross-Linked Star Polymer Model Networks Containing a Novel, Silicon-Based, Hydrolyzable Cross-Linker
Themistou E and Patrickios CS.
Macromolecules, 37(18), 6734-6743 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service