Skip to Content
Merck
All Photos(1)

Key Documents

799084

Sigma-Aldrich

Graphene nanoplatelets

powder, avg. no. of layers, 5 ‑ 7

Synonym(s):

GNP powder, Graphene powder

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
Molecular Weight:
12.01
EC Number:
UNSPSC Code:
12352103

description

Surfactant type: Anionic Surfactant

form

powder

feature

avg. no. of layers 5 ‑ 7

composition

Graphene nanoplates as produced

sheet resistance

10 (+/-5) Ω/sq (for a 25 μm film)

Looking for similar products? Visit Product Comparison Guide

General description

Raman D/G and D/D′: 0.28 and 5.0 (The 2D band shape indicates the presence of few-layer graphene flakes with an average of 5-7 atomic layers)

Application

  • Graphene (nano)composite materials
  • Conductive coatings
  • Conductive Inks
  • Energy Storage

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Xiaofeng Liu et al.
Advanced materials (Deerfield Beach, Fla.), 25(43), 6284-6290 (2013-08-22)
Inspired by black powder explosion, a mixture of sugar and nitrate is diluted with inert molten salt, such that their rapid reaction is moderated, allowing capturing of the intermediacy product: highly porous N-doped graphene nanoplates. The methodology is extended to
Metin Uz et al.
ACS applied materials & interfaces, 12(11), 13529-13539 (2020-02-23)
In this study, a simple microfluidic method, which can be universally applied to different rigid or flexible substrates, was developed to fabricate high-resolution, conductive, two-dimensional and three-dimensional microstructured graphene-based electronic circuits. The method involves controlled and selective filling of microchannels
Electrical conductivity maps in graphene nanoplatelet/silicon nitride composites using conducting scanning force microscopy
Ramirez, C.; Garzon, L.; Miranzo, P.; et al.
Carbon, 49(12), 3873-3880 (2011)

Articles

3D printing is a type of additive manufacturing that can be used to rapidly fabricate components with highly customizable geometries.

Graphene is a unique two-dimensional (2D) structure of monolayer carbon atoms packed into a dense honeycomb crystal that has attracted great interest due to its diverse and fascinating properties.

Graphene's unique properties spark interdisciplinary interest; its honeycomb structure offers electrical, optical, and mechanical marvels.

Since its discovery little more than a decade ago,1 the two-dimensional (2D) allotrope of carbon—graphene—has been the subject of intense multidisciplinary research efforts.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service