Skip to Content
Merck
All Photos(1)

Key Documents

797774

Sigma-Aldrich

Graphene nanoribbons

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C
Molecular Weight:
12.01
EC Number:
UNSPSC Code:
12352103
NACRES:
NA.23

Assay

≥90.0% carbon basis (TGA)

form

powder

L × W

2-15 μm × 40-250 nm

density

2.2745 g/mL (He gas method)

bulk density

0.0970 g/mL (Mercury Porosimetry)

Looking for similar products? Visit Product Comparison Guide

General description

Produced by unzipping multi-walled carbon nanotubes by potassium intercalation.

Application

  • Composites.
  • Conductive inks.
  • Electrodes for LiB.
  • Energy storage & harvesting applications.
  • Bio-medical applications.
  • Preferred dispersing organic solvents: pyrrolidones and chlorinated solvents.
  • Less preferred dispersing organic solvents: cyclohexanone and γ-butyrolactone.
  • Aqueous dispersions are possible at 0.1mg/mL with triton-X-100, sodium cholate and deoxycholate and cellulose-based surfactants.

Analysis Note

Raman Spectroscopic Data:
ID/IG = 0.65±0.07
I2D/IG = 0.74±0.03
2D FWHM = 63 cm-1

Legal Information

Graphene Nanoribbon Composites and Methods of Making the Same, WO/2012/112435 A1.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 2 - STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

nwg


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Changsheng Xiang et al.
ACS nano, 7(11), 10380-10386 (2013-10-10)
A thermoplastic polyurethane (TPU) composite film containing hexadecyl-functionalized low-defect graphene nanoribbons (HD-GNRs) was produced by solution casting. The HD-GNRs were well distributed within the polyurethane matrix, leading to phase separation of the TPU. Nitrogen gas effective diffusivity of TPU was
High Throughput Preparation of Large Area Transparent Electrodes Using Non-Functionalized Graphene Naroribbons.
Zhu Y, et al.
Chemistry of Materials, 23, 935-939 (2011)
Dmitry V Kosynkin et al.
ACS nano, 5(2), 968-974 (2011-01-06)
Here we demonstrate that graphene nanoribbons (GNRs) free of oxidized surfaces can be prepared in large batches and 100% yield by splitting multiwalled carbon nanotubes (MWCNTs) with potassium vapor. If desired, exfoliation is attainable in a subsequent step using chlorosulfonic
Bostjan Genorio et al.
ACS nano, 6(5), 4231-4240 (2012-04-04)
A cost-effective and potentially industrially scalable, in situ functionalization procedure for preparation of soluble graphene nanoribbon (GNRs) from commercially available carbon nanotubes is presented. The physical characteristics of the functionalized product were determined using SEM, evolved gas analysis, X-ray diffraction
Lei Li et al.
ACS applied materials & interfaces, 5(14), 6622-6627 (2013-06-25)
A facile and cost-effective approach to the fabrication of a nanocomposite material of polyaniline (PANI) and graphene nanoribbons (GNRs) has been developed. The morphology of the composite was characterized by scanning electron microscopy, transmission electron microscopy, X-ray photoelectron microscopy, and

Articles

Graphene is a unique two-dimensional (2D) structure of monolayer carbon atoms packed into a dense honeycomb crystal that has attracted great interest due to its diverse and fascinating properties.

Graphene's unique properties spark interdisciplinary interest; its honeycomb structure offers electrical, optical, and mechanical marvels.

Since its discovery little more than a decade ago,1 the two-dimensional (2D) allotrope of carbon—graphene—has been the subject of intense multidisciplinary research efforts.

Graphene nanoribbons (GNRs) are quasi-one-dimensional narrow strips of graphene comprised of sp2-hybridized carbon atoms arranged into hexagonal honeycomb lattice configurations.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service