Skip to Content
Merck
All Photos(2)

Key Documents

796670

Sigma-Aldrich

2,2,3,3-Tetramethylsuccinonitrile

Synonym(s):

2,2,3,3-Tetramethylbutanedinitrile, 2,3-Dicyano-2,3-dimethylbutane, Tetramethylsuccinic acid dinitrile, Tetramethylsuccinodinitrile

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$90.10

$90.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$90.10

About This Item

Empirical Formula (Hill Notation):
C8H12N2
CAS Number:
Molecular Weight:
136.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$90.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

solid

Quality Level

functional group

nitrile

SMILES string

N#CC(C)(C)C(C)(C)C#N

InChI

1S/C8H12N2/c1-7(2,5-9)8(3,4)6-10/h1-4H3

InChI key

ZVQXQPNJHRNGID-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,2,3,3-Tetramethylsuccinonitrile can be used as a reactant to synthesize:
  • 3,4-Dihydro-3H-pyrrol-2-imines by reacting with aryl lithium species in the presence of TMSCl.[1]
  • Phenyl substituted metal-free fused tetraazachlorins by condensation reaction with substituted phthalonitrile derivatives in the presence of InCl3.[2]

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - STOT RE 2 Inhalation - STOT SE 2 Inhalation

Target Organs

Kidney,Liver, Nervous system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

3, 4-Dihydro-3H-pyrrol-2-imines as Conformationally Restrained 1, 3-Diazabutadienes: Synthesis, Structural Properties and Protonation
Janich S, et al.
European Journal of Organic Chemistry, 2077-2087 (2009)
An efficient synthesis of metal-free tetraazachlorins via indium complexes
Makarova EA, et al.
Journal of Porphyrins and Phthalocyanines, 17(08n09), 785-790 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service