Skip to Content
Merck
All Photos(2)

Key Documents

794252

Sigma-Aldrich

2,7-Dimethyl-9(10H)-acridinone

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$435.00

$435.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$435.00

About This Item

Empirical Formula (Hill Notation):
C15H13NO
Molecular Weight:
223.27
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$435.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Quality Level

Assay

97%

form

solid

SMILES string

CC1=CC2=C(NC(C=CC(C)=C3)=C3C2=O)C=C1

InChI

1S/C15H13NO/c1-9-3-5-13-11(7-9)15(17)12-8-10(2)4-6-14(12)16-13/h3-8H,1-2H3,(H,16,17)

InChI key

IBEGAUGEWHXHQH-UHFFFAOYSA-N

Application

Product was reported as an intermediate toward the synthesis of acridine-containing 1, 2-dioxetanes which have been utilized as thermochemiluminescence labels in bioanalysis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Nicewicz lab focuses on reactive cation radical species for selective reactions, advancing synthesis methodologies for various functional groups.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service