Skip to Content
Merck
All Photos(1)

Key Documents

794198

Sigma-Aldrich

Neopentyl-tBu2P Pd G2

Synonym(s):

Chloro[(di-tert-butylneopentylphosphine)-2-(2-aminobiphenyl)]palladium(II)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H39ClNPPd
CAS Number:
Molecular Weight:
526.43
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

form

solid

feature

generation 2

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings

functional group

phosphine

SMILES string

Cl[Pd-]1C2=CC=CC=C2C3=C([NH2+]1)C=CC=C3.CC(C)(C)P(C(C)(C)C)CC(C)(C)C

InChI

1S/C13H29P.C12H10N.ClH.Pd/c1-11(2,3)10-14(12(4,5)6)13(7,8)9;13-12-9-5-4-8-11(12)10-6-2-1-3-7-10;;/h10H2,1-9H3;1-6,8-9H,13H2;1H;/q;;;+1/p-1

InChI key

DQUVXGBXLUAOQM-UHFFFAOYSA-M

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service