Skip to Content
Merck
All Photos(2)

Key Documents

792152

Sigma-Aldrich

4-Methylpiperidine solution

suitable for peptide synthesis, 20 % (w/w) in DMF

Synonym(s):

4-Methylpiperidine solution

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
$226.00
2 L
$824.00
4 L
$944.00

$226.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
500 ML
$226.00
2 L
$824.00
4 L
$944.00

About This Item

Empirical Formula (Hill Notation):
C6H13N
CAS Number:
Molecular Weight:
99.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$226.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Assay

≥95.5%

Quality Level

form

liquid

reaction suitability

reagent type: reductant

concentration

20 % (w/w) in DMF

refractive index

n20/D 1.433

density

0.920 g/mL at 25 °C

application(s)

peptide synthesis

SMILES string

N1CCC(CC1)C

InChI

1S/C6H13N/c1-6-2-4-7-5-3-6/h6-7H,2-5H2,1H3

InChI key

UZOFELREXGAFOI-UHFFFAOYSA-N

Application

4-Methylpiperidine solution (20 % in DMF) can be used:
  • In the deprotection of F-moc protecting group which is attached to free amine.[1]
  • In the determination of resin-bound amino acid by UV-Fmoc-quantitation.[2]

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 3 - Repr. 1B - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

98.8 °F - closed cup

Flash Point(C)

37.1 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Design, solid-phase synthesis and evaluation of enterobactin analogs for iron delivery into the human pathogen Campylobacter jejuni
Zamora CY, et al.
Bioorganic & Medicinal Chemistry, 26(19), 5314-5321 (2018)
A sensitive and practical colorimetric test for polymer-supported hydroxyl and thiol groups
Caroen J and Van der EJ
Tetrahedron Letters, 50(1), 41-44 (2009)
Biljana Mojsoska et al.
Antimicrobial agents and chemotherapy, 59(7), 4112-4120 (2015-05-06)
The constant emergence of new bacterial strains that resist the effectiveness of marketed antimicrobials has led to an urgent demand for and intensive research on new classes of compounds to combat bacterial infections. Antimicrobial peptoids comprise one group of potential
Leopold Kong et al.
Journal of molecular biology, 427(16), 2617-2628 (2015-07-03)
Hepatitis C virus (HCV) is a positive-strand RNA virus within the Flaviviridae family. The viral "spike" of HCV is formed by two envelope glycoproteins, E1 and E2, which together mediate viral entry by engaging host receptors and undergoing conformational changes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service