Skip to Content
Merck
All Photos(2)

Key Documents

79179

Sigma-Aldrich

Nitromethyl phenyl sulfone

≥99.0% (T)

Synonym(s):

(Nitromethylsulfonyl)benzene, (Phenylsulfonyl)nitromethane, Benzenesulfonylnitromethane

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$327.00

$327.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$327.00

About This Item

Linear Formula:
C6H5SO2CH2NO2
CAS Number:
Molecular Weight:
201.20
Beilstein:
1876502
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$327.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥99.0% (T)

form

crystals

mp

78-80 °C

functional group

amine
nitro
sulfone

SMILES string

[O-][N+](=O)CS(=O)(=O)c1ccccc1

InChI

1S/C7H7NO4S/c9-8(10)6-13(11,12)7-4-2-1-3-5-7/h1-5H,6H2

InChI key

ADKLJTCVZHUANG-UHFFFAOYSA-N

Other Notes

Versatile acidic analogue of nitromethane[1]; Used for preparing phenylsulfonyl nitrile oxide, a reagent for syn-cyanohydroxylations of olefins[2][3]; Cycloadditions with olefins[4]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E.E. Knaus
The Journal of Organic Chemistry, 49, 123-123 (1984)
P.A. Wade et al.
The Journal of Organic Chemistry, 46, 5425-5425 (1981)
P.A. Wade et al.
The Journal of Organic Chemistry, 46, 765-765 (1981)
P.A. Wade et al.
The Journal of Organic Chemistry, 49, 4595-4595 (1984)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service