Skip to Content
Merck
All Photos(1)

Key Documents

79135

Sigma-Aldrich

Phenylmercury nitrate

basic, ≥97.0% (RT)

Synonym(s):

Phenylmercuric nitrate, basic

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
$211.00

$211.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
50 G
$211.00

About This Item

Linear Formula:
C6H5HgNO3·C6H5HgOH
CAS Number:
Molecular Weight:
634.40
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$211.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Assay

≥97.0% (RT)

form

powder

mp

175-185 °C (dec.) (lit.)

SMILES string

O[Hg]c1ccccc1.[O-][N+](=O)O[Hg]c2ccccc2

InChI

1S/2C6H5.2Hg.NO3.H2O/c2*1-2-4-6-5-3-1;;;2-1(3)4;/h2*1-5H;;;;1H2/q;;2*+1;-1;/p-1

InChI key

DQZCXZOUXZTNCU-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

It is used as a preservative for parenteral injections.[1]

Disclaimer

As part of the corporation global plan to support the Minamata Convention eliminating mercury-containing compounds, as of January 2021 customers will be required to provide an End-Use Declaration. The sales of thimerosal will be restricted for approved applications only, such as preservative in vaccine manufacturing, preservative in eye cosmetics, research use within academia and as reference material only. The product is not intended for use as a biocide under global biocide regulations, including but not limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act, European Biocidal Products Regulation, Canada’s Pest Management Regulatory Agency, Turkey’s Biocidal Products Regulation, Korea’s Consumer Chemical Products and Biocide Safety Management Act (K-BPR) and others.
For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - STOT RE 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Disinfection
Bentley's Textbook of Pharmaceutics (2012)

Questions

  1. If one is allergic to metal could phenyl mercuric nitrate causr an allergic reaction when used in eye drops as a preservative?

    1 answer
    1. Unless otherwise stated, this product is intended for research use only. It is not suitable for any other purpose, which includes, but is not limited to, unauthorized commercial use, in vitro diagnostic use, or therapeutic use in humans or animals

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service