Skip to Content
Merck
All Photos(1)

Key Documents

791164

Sigma-Aldrich

Polystyrene, amine terminated

average Mn 5,000, PDI ≤1.2

Synonym(s):

PS NH2, amino PS, aminopolystyrene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C8H8)nCH5N
UNSPSC Code:
12352100
NACRES:
NA.23

form

powder

mol wt

average Mn 5,000

mp

300 °C

PDI

≤1.2

Looking for similar products? Visit Product Comparison Guide

Application

Functional polystyrene with amine termination has been used as a macromolecular surfactant; capable of conjugation with enzymes; proteins and lipases. They are also known to form well-defined assemblies in aqueous solution.

Legal Information

Sold for research purposes only: See sigma-aldrich.com/raftlicense. Patents 7; 714; 075; 7; 250; 479; 7; 666; 962; 6; 642; 318; 6; 747; 111; W2010/8356.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dirks; A. J.;
Chemical Communications (Cambridge, England), 4172-4174 (2005)

Articles

Biomaterials science integrates smart materials into biological research, requiring a deep understanding of biological systems.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service