Skip to Content
Merck
All Photos(1)

Documents

790621

Sigma-Aldrich

Methyl 2,3,4-tri-O-acetyl-6-O-trityl-α-D-galactopyranoside

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H34O9
CAS Number:
Molecular Weight:
562.61
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

97%

optical activity

[α]22/D +55°, c = 1% in chloroform

mp

177-182 °C

storage temp.

−20°C

SMILES string

CC(O[C@H]1[C@@H](COC(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4)O[C@H](OC)[C@H](OC(C)=O)[C@H]1OC(C)=O)=O

InChI

1S/C32H34O9/c1-21(33)38-28-27(41-31(36-4)30(40-23(3)35)29(28)39-22(2)34)20-37-32(24-14-8-5-9-15-24,25-16-10-6-11-17-25)26-18-12-7-13-19-26/h5-19,27-31H,20H2,1-4H3/t27-,28+,29+,30-,31+/m1/s1

InChI key

CEIHIENXWVDRTC-NBCLCUQJSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service