Skip to Content
Merck
All Photos(1)

Key Documents

790397

Sigma-Aldrich

Potassium 2-bromo-4-(trifluoromethyl)pyridine-3-trifluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H2BBrF6KN
Molecular Weight:
331.89
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

SMILES string

BrC1=C([B-](F)(F)F)C(C(F)(F)F)=CC=N1.[K+]

InChI

1S/C6H2BBrF6N.K/c8-5-4(7(12,13)14)3(1-2-15-5)6(9,10)11;/h1-2H;/q-1;+1

InChI key

WGDRKFJAJWNHQI-UHFFFAOYSA-N

Application

An organotrifluoroborate involved in:
  • Nickel-catalyzed cross-coupling reactions and C-O activation
  • Suzuki cross-coupling,
  • Copper-catalyzed cross-coupling reactions
  • Stereoselective Mukaiyama aldol reactions

Organotrifluoroborates as versatile and stable boronic acid surrogates.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of 5-aryl- and 5-heteroaryl-7-carboxyl-8-hydroxyquinaldines through Suzuki cross-coupling reaction with potassium organotrifluoroborates
Synthesis, 4, 619-630 (2010)
Gary A Molander et al.
Organic letters, 12(18), 4022-4025 (2010-08-19)
A general method based on nickel-catalyzed C-O activation of various phenol derivatives with potassium (hetero)aryltrifluoroborates has been developed. A large number of heterobiaryls can be easily obtained with yields up to 99% using methanesulfonate cross-coupling partners.
Copper catalyzed cross-coupling reactions of diaryl ditellurides with potassium aryltrifluoroborate salts
Journal of the Brazilian Chemical Society, 20, 988-992 (2009)
J Michael Ellis et al.
The Journal of organic chemistry, 73(22), 9151-9154 (2008-10-16)
A new synthesis of 3,3'-bioxindoles is reported that is well suited for the preparation of unsymmetrical structures. In the key step, 3-hydroxy-3,3'-bioxindoles are constructed by Mukaiyama aldol reaction of 2-siloxyindoles with isatins. These tertiary carbinols are formed in high diastereoselectivities
Gary A Molander et al.
The Journal of organic chemistry, 74(3), 973-980 (2008-12-25)
A wide variety of bench-stable potassium heteroaryltrifluoroborates were prepared, and general reaction conditions were developed for their cross-coupling to aryl and heteroaryl halides. The cross-coupled products were obtained in good to excellent yields. This method represents an efficient and facile

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service