78153
N-Phenylanthranilic acid
technical, ≥95% (T)
Synonym(s):
2-(Phenylamino)benzoic acid, Diphenylamine-2-carboxylic acid, DpC
About This Item
Recommended Products
grade
technical
Assay
≥95% (T)
form
powder
mp
182-185 °C (lit.)
182-185 °C
application(s)
peptide synthesis
SMILES string
OC(=O)c1ccccc1Nc2ccccc2
InChI
1S/C13H11NO2/c15-13(16)11-8-4-5-9-12(11)14-10-6-2-1-3-7-10/h1-9,14H,(H,15,16)
InChI key
ZWJINEZUASEZBH-UHFFFAOYSA-N
Gene Information
human ... TTR(7276)
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service